Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3346

Sigma-Aldrich

3-PEHPC

≥95% (HPLC)

Synonym(s):

α-Hydroxy-α-phosphono-3-pyridinepropanoic acid;, 2-Hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid, NE10790

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO6P
CAS Number:
Molecular Weight:
247.14
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

3-PEHPC (NE 10790), a phosphonocarboxylate analogue of the bisphosphonate risedronate, is a potent and selective inhibitor of geranylgeranyl transferase II (GGTase II).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caihong Wang et al.
Autophagy, 15(3), 527-542 (2018-10-20)
50% of Caucasians carry a Thr300Ala variant (T300A) in the protein encoded by the macroautophagy/autophagy gene ATG16L1. Here, we show that the T300A variant confers protection against urinary tract infections (UTIs), the most common infectious disease in women. Using knockin
Beth Binnington et al.
Glycobiology, 26(2), 166-180 (2015-09-26)
Statins, which specifically inhibit HMG Co-A reductase, the rate-limiting step of cholesterol biosynthesis, are widely prescribed to reduce serum cholesterol and cardiac risk, but many other effects are seen. We now show an effect of these drugs to induce profound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service