Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3370

Sigma-Aldrich

BI605906

≥98% (HPLC)

Synonym(s):

3-Amino-4-(1,1-difluoro-propyl)-6-(4-methanesulfonyl-piperidin-1-yl)-thieno[2,3-b]pyridine-2-carboxylic acid amide, 3-Amino-4-(1,1-difluoropropyl)-6-[4-(methylsulfonyl)-1-piperidinyl]-thieno[2,3-b]pyridine-2-carboxamide, BI 605906, BI-605906

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22F2N4O3S2
CAS Number:
Molecular Weight:
432.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCC(F)(C1=C2C(SC(C(N)=O)=C2N)=NC(N3CCC(CC3)S(C)(=O)=O)=C1)F

Biochem/physiol Actions

BI605906 is a selective, ATP-competitive IκB kinase β (IKK2, IKKβ, IKK-beta) inhibitor (IC50 = 380 nM, [ATP] = 0.1 mM) with no potency toward 100 other kinases, including IKKα, IKKε, TBK1, and IGF1R (IC50 ≥7.6 µM). BI605906 (10 µM) effectively blocks IKK-β-dependent phosphorylation of IKKε and TBK1 in IKKα-/- MEFs upon IL-1α (5 ng/mL) induction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristopher Clark et al.
The Biochemical journal, 434(1), 93-104 (2010-12-09)
Members of the IKK {IκB [inhibitor of NF-κB (nuclear factor κB)] kinase} family play a central role in innate immunity by inducing NF-κB- and IRF [IFN (interferon) regulatory factor]-dependent gene transcription programmes required for the production of pro-inflammatory cytokines and
Amy R Cameron et al.
Circulation research, 119(5), 652-665 (2016-07-16)
The diabetes mellitus drug metformin is under investigation in cardiovascular disease, but the molecular mechanisms underlying possible benefits are poorly understood. Here, we have studied anti-inflammatory effects of the drug and their relationship to antihyperglycemic properties. In primary hepatocytes from
Raid B Nisr et al.
Cellular and molecular life sciences : CMLS, 76(24), 4887-4904 (2019-05-19)
Sustained nutrient (fuel) excess, as occurs during obesity and diabetes, has been linked to increased inflammation, impaired mitochondrial homeostasis, lipotoxicity, and insulin resistance in skeletal muscle. Precisely how mitochondrial dysfunction is initiated and whether it contributes to insulin resistance in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service