Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3288

Sigma-Aldrich

WX8

≥98% (HPLC)

Synonym(s):

1H-Indole-3-carbaldehyde [4-anilino-6-(4-morpholinyl)-1,3,5-triazin-2-yl]hydrazone, 1H-Indole-3-carboxaldehyde, [4-(4-morpholinyl)-6-(phenylamino)-1,3,5-triazin-2-yl]hydrazone, Ro 91-4714, Ro-91-4714

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N8O
CAS Number:
Molecular Weight:
414.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C1(NC2=CC=CC=C2)=NC(N/N=C/C3=CNC4=C3C=CC=C4)=NC(N5CCOCC5)=N1

InChI key

NJIKLALXAPYTCW-BUVRLJJBSA-N

Biochem/physiol Actions

Selective phosphoinositide kinase PIKfyve inhibitor with higher antiproliferation potency in A375 cultures than YM201636, chloroquine or hydroxychloroquine.
WX8 (Ro 91-4714) is a selective phosphoinositide kinase PIKfyve inhibitor that disrupts lysosome fission via tubulation, lysosomal trafficking, and heterotypic lysosomes-autophagosomes fusion (0.1-1 µM; U2OS), but not homotypic lysosome fusion. WX8 exhibits cancer-selective antiproliferation activity (IC50 = 48 nM/A375, 200 nM/U2OS; IC50 >10 µM/non-cancer 293T & HFF) with a higher potency than the PIKfyve inhibitor YM201636, or the lysosomal inhibitors chloroquine & hydroxychloroquine (A375 IC50 = 119 nM, 1.7 µM, 1.9 µM, repectively).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajit Roy et al.
Molecular oncology, 18(4), 988-1011 (2024-02-28)
Inhibitors specifically targeting the 1-phosphatidylinositol 3-phosphate 5-kinase (PIKFYVE) disrupt lysosome homeostasis, thereby selectively terminating autophagy-dependent human cancer cells in vivo as well as in vitro without harming the viability of nonmalignant cells. To elucidate the mechanism by which PIKFYVE inhibition
Ajit Roy et al.
Autophagy, 19(9), 2464-2484 (2023-02-22)
Although PIKFYVE phosphoinositide kinase inhibitors can selectively eliminate PIKFYVE-dependent human cancer cells in vitro and in vivo, the basis for this selectivity has remained elusive. Here we show that the sensitivity of cells to the PIKFYVE inhibitor WX8 is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service