Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3271

Sigma-Aldrich

Etoricoxib

≥95% (HPLC)

Synonym(s):

5-Chloro-3-[4-(methylsulfonyl)phenyl]-2-(2-methyl-5-pyridinyl)pyridine, 5-Chloro-6′-methyl-3-[4-(methylsulfonyl)phenyl]-2,3′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15ClN2O2S
CAS Number:
Molecular Weight:
358.84
Beilstein:
8073797
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=NC=C(C2=C(C3=CC=C(S(C)(=O)=O)C=C3)C=C(Cl)C=N2)C=C1

InChI

1S/C18H15ClN2O2S/c1-12-3-4-14(10-20-12)18-17(9-15(19)11-21-18)13-5-7-16(8-6-13)24(2,22)23/h3-11H,1-2H3

InChI key

MNJVRJDLRVPLFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Riendeau et al.
The Journal of pharmacology and experimental therapeutics, 296(2), 558-566 (2001-02-13)
We report here the preclinical profile of etoricoxib (MK-0663) [5-chloro-2-(6-methylpyridin-3-yl)-3-(4-methylsulfonylphenyl) pyridine], a novel orally active agent that selectively inhibits cyclooxygenase-2 (COX-2), that has been developed for high selectivity in vitro using whole blood assays and sensitive COX-1 enzyme assays at
Sebastián A Cuesta et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-20)
In this review, a timeline starting at the willow bark and ending in the latest discoveries of analgesic and anti-inflammatory drugs will be discussed. Furthermore, the chemical features of the different small organic molecules that have been used in pain
Lalita Tanwar et al.
Asian Pacific journal of cancer prevention : APJCP, 11(5), 1329-1333 (2011-01-05)
In the present study, we assessed effects of etoricoxib, a non-steroidal anti-inflammatory drug, on proliferation and apoptosis in 1,2-dimethylhydrazine dihydrochloride (DMH) induced colon lesion development. Male SD rats were divided into four groups: Group 1 controls receiving the vehicle treatment;

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service