Skip to Content
MilliporeSigma

D0638

Dibucaine hydrochloride

analytical standard, ≥99%

Synonym(s):

2-Butoxy-N-(2-diethylaminoethyl)-4-quinolinecarboxamide hydrochloride, Cinchocaine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H29N3O2 · HCl
CAS Number:
Molecular Weight:
379.92
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-498-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥99%

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

99-101 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1

InChI

1S/C20H29N3O2.ClH/c1-4-7-14-25-19-15-17(16-10-8-9-11-18(16)22-19)20(24)21-12-13-23(5-2)6-3;/h8-11,15H,4-7,12-14H2,1-3H3,(H,21,24);1H

InChI key

IVHBBMHQKZBJEU-UHFFFAOYSA-N

Gene Information

General description

Dibucaine hydrochloride is a quinolone derivative, which can be prepared using isatin as the starting material followed by a series of reactions. It is a local anesthetic agent.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potent, long-acting local anesthetic


Still not finding the right product?

Explore all of our products under Dibucaine hydrochloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library