Skip to Content
MilliporeSigma

Skip To

SML3258

3-Methyl-5-(2,2,3-trimethyl-3-cyclopentenyl)pentan-2-ol

≥90% (GC)

Synonym(s):

3-Methyl-5-(2,2,3-trimethylcyclopent-3-en-1-yl)pentan-2-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H26O
CAS Number:
Molecular Weight:
210.36
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥90% (GC)
Form:
oil
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (GC)

form

oil

color

white to beige

storage temp.

−20°C

SMILES string

CC1=CCC(CCC(C)C(C)O)C1(C)C

InChI

1S/C14H26O/c1-10(12(3)15)6-8-13-9-7-11(2)14(13,4)5/h7,10,12-13,15H,6,8-9H2,1-5H3

InChI key

NGYMOTOXXHCHOC-UHFFFAOYSA-N

Biochem/physiol Actions

Agonist of olfactory receptor OR2AT4
Sandalore is an agonist of olfactory receptor OR2AT4, a G-protein-coupled receptor (GPCR) also expressed in human primary keratinocytes. Sandalore was shown to enhance hair growth in human scalp hair follicles by stimulating OR2AT4, resulting in decreased apoptosis and increased production of the anagen-prolonging growth factor IGF-1. Sandalore had previously been shown to increase wound healing. Sandalore was also recently used to study OR2AT4 found in the human chronic myelogenous leukemia (CML) cell line K562 and in white blood cells of clinically diagnosed acute myeloid leukemia (AML) patients.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6153074959811584
description

≥90% (GC)

description

98 atom % D

description

analytical standard

description

≥95.0% (sum of enantiomers, GC)

assay

≥90% (GC)

assay

-

assay

≥95.0% (sum of enantiomers, GC)

assay

≥95.0% (sum of enantiomers, GC)

form

oil

form

-

form

-

form

-

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

color

white to beige

color

-

color

-

color

-


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters