Skip to Content
MilliporeSigma
All Photos(2)

Documents

49598

Supelco

(−)-Terpinen-4-ol

analytical standard

Synonym(s):

(R)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (R)-p-Menth-1-en-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]/D -25±6°, neat

optical purity

enantiomeric ratio: ~2:1

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.934 g/mL at 20 °C

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@@]1(O)CCC(C)=CC1

InChI

1S/C10H18O/c1-8(2)10(11)6-4-9(3)5-7-10/h4,8,11H,5-7H2,1-3H3/t10-/m0/s1

InChI key

WRYLYDPHFGVWKC-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Lan et al.
Journal of Zhejiang University. Science. B, 15(11), 940-952 (2014-11-05)
Our previous studies had confirmed that the essential oil from Zanthoxylum bungeanum Maxim. (Z. bungeanum oil) could effectively enhance the percutaneous permeation of drug molecules as a natural transdermal penetration enhancer. The aim of the present study is to investigate
Raimunda Sâmia Nogueira Brilhante et al.
Medical mycology, 53(2), 93-98 (2014-12-30)
Sporotrichosis is a subacute or chronic subcutaneous infection, caused by the fungus Sporothrix schenkii complex, occurring in human and animal tissues. Potassium iodide and itraconazole have been used as effective therapy for first-choice treatment, while amphotericin B may be indicated
Seon-Mi Seo et al.
Pesticide biochemistry and physiology, 113, 55-61 (2014-07-24)
This study investigated the fumigant toxicity of 4 Asteraceae plant essential oils and their constituents against the Japanese termite Reticulitermes speratus Kolbe. Fumigant toxicity varied with plant essential oils or constituents, exposure time, and concentration. Among the tested essential oils
Christian Zorzetto et al.
Fitoterapia, 100, 95-109 (2014-12-03)
In the present work we carried out a phytochemical and biological investigation on three Hypericum species, i.e. Hypericum reflexum, Hypericum canariense and Hypericum grandifolium, from the Canary Islands where they are traditionally used as diuretic, wound healing, vermifuge, sedative and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service