Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2883

Sigma-Aldrich

Paquinimod

≥98% (HPLC), powder, immunomodulator

Synonym(s):

ABR 215757, ABR 25757, ABR-215757, ABR-25757, ABR215757, ABR25757, N,5-Diethyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-N-phenyl-3-quinolinecarboxamide, N-Ethyl-N-phenyl-1,2-dihydro-5-ethyl-4-hydroxy-1-methyl-2-oxoquinoline-3-carboxamide, N-Ethyl-N-phenyl-5-ethyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxoquinoline-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O3
CAS Number:
Molecular Weight:
350.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Paquinimod, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Paquinimod (ABR-215757) is an orally active quinoline-3-carboxamide (Q substance) class immunomodulator that targets S100A9 (Calgranulin B; MRP14) via direct binding and blocks its interaction with receptor for advanced glycation end products (RAGE) and Toll-like receptor 4 (IC50 = 26 μM & 23 μM against 100 nM hS100A9 from binding immobilized hRAGE & hTLR4/MD2, respectively). Paquinimod in vivo efficacy is demonstrated in murine models of autoimmune/inflammatory diseases, including liver fibrosis, collagen-induced osteoarthritis, peritonitis, EAE, type 1 diabetes, lupus (0.04-25 mg/kg/day p.o.), and atherosclerosis (10 mg/kg i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stig Jönsson et al.
Journal of medicinal chemistry, 47(8), 2075-2088 (2004-04-02)
Roquinimex-related 3-quinolinecarboxamide derivatives were prepared and evaluated for treatment of autoimmune disorders. The compounds were tested in mice for their inhibitory effects on disease development in the acute experimental autoimmune encephalomyelitis model and selected compounds in the beagle dog for
Hans Carlsten et al.
International immunopharmacology, 4(12), 1515-1523 (2004-09-08)
Autoimmune, lupus-prone MRL lpr/lpr mice were treated orally with oxo-quinoline-3-carboxamide (ABR-25757), a newly developed immunomodulator. Treatment was initiated in one set of experiment at the age of 10 weeks, before the onset of clinically apparent disease, and in another set
Hong Gong et al.
Journal of neuroinflammation, 15(1), 252-252 (2018-09-06)
Depression is one of the most common mental disorders characterized mainly by low mood and loss of interest or pleasure. About a third of patients with depression do not respond to classic antidepressant treatments. Recent evidence suggests that Mrp8/14 (myeloid-related
Nina Fransén Pettersson et al.
PloS one, 13(9), e0203228-e0203228 (2018-09-06)
Quinoline-3-carboxamides (Q substances) are small molecule compounds with anti-inflammatory properties. In this study, we used one of these substances, Paquinimod, to treat a novel model for chronic liver inflammation and liver fibrosis, the NOD-Inflammation Fibrosis (N-IF) mouse. We show that
Ling Yan et al.
Atherosclerosis, 228(1), 69-79 (2013-03-19)
There is an emerging widespread interest in the role of damage-associated molecular pattern molecules (DAMP) S100A8, S100A9 and S100A12 in cardiovascular and other diseases. In this study we tested the efficacy of ABR-215757, a S100 protein binding immuno-modulatory compound to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service