Skip to Content
MilliporeSigma

5341

Progesterone

≥98% (HPLC), Steroid hormone, crystalline solid

Synonym(s):

Progesterone, Δ⁴-Pregnene-3,20-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
crystalline solid
Quality level:
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Progesterone, Steroid hormone secreted by the corpus luteum during the latter half of the menstrual cycle.

description

RTECS - TW0175000
Merck USA index - 14, 7773

Quality Level

assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

O=C1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)C(=O)C)(CC3)C)CCC2=C1)C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Steroid hormone secreted by the corpus luteum during the latter half of the menstrual cycle. Binds to the progesterone receptor in the nucleus to influence transcription. Used as a contraceptive agent and in the treatment of uterine disorders.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Binds to the progesterone receptor in the nucleus to influence transcription
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, refrigerate (4°C). Ethanol stock solutions are stable for up to 6 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
155683-5G04061833371640
5341-25GM04055977269369

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service