Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2341

Sigma-Aldrich

Nω-Propyl-L-Arginine hydrochloride

≥98% (HPLC)

Synonym(s):

L-NPA, N(ω)-Propyl-L-arginine, N(w)-Propyl-L-arginine, N-ω-Propyl-L-arginine, N-Omega-propyl-L-Arginine, N5-[Imino(propylamino)methyl]-L-ornithine, NG-Propyl-L-arginine, NPA, NPLA, Nomega-Propyl-L-Arginine, PLA, p-ARG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20N4O2 · xHCl
CAS Number:
Molecular Weight:
216.28 (free base basis)
MDL number:
UNSPSC Code:
12352209

Assay

≥98% (HPLC)

form

film

storage condition

desiccated

color

colorless

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCNC(NCCC[C@H](N)C(O)=O)=N

InChI

1S/C9H20N4O2/c1-2-5-12-9(11)13-6-3-4-7(10)8(14)15/h7H,2-6,10H2,1H3,(H,14,15)(H3,11,12,13)/t7-/m0/s1

InChI key

AOMXURITGZJPKB-ZETCQYMHSA-N

Biochem/physiol Actions

Nω-Propyl-L-arginine (NPA; NPLA; p-ARG; L-PNA) is a substrate-competitive, reversible, potent and selective neuronal nitric oxide synthase (nNOS; bNOS) inhibitor (Ki = 57 nM/bovine nNOS, 8.5 μM/bovine eNOS, 180 μM/murine iNOS) widely used for probing nNOS-dependent physiological and pathological processes both in cultures and in animals in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiting Zhang et al.
Neurobiology of disease, 110, 122-132 (2017-12-06)
Germinal matrix hemorrhage is induced by stereotaxic injection of collagenase into the germinal matrix of P7 Sprague-Dawley rats. Hemoglobin assay, western blot, immunofluorescence and neurobehavioral tests were used to test the effects of BLVRA on hematoma resolution and anti-inflammatory response.
Qing-Bo Lu et al.
International journal of molecular sciences, 19(1), 59-59 (2017-12-28)
This study was conducted to explore the hypothesis that the endogenous superoxide anions (O₂-) and nitric oxide (NO) system of the paraventricular nucleus (PVN) regulates the cardiac sympathetic afferent reflex (CSAR) contributing to sympathoexcitation in obese rats induced by a
Al Muktafi Sadi et al.
Clinical science (London, England : 1979), 132(6), 641-654 (2018-03-01)
Background: Heart failure (HF) is associated with reduced expression of plasma membrane Ca2+-ATPase 4 (PMCA4). Cardiac-specific overexpression of human PMCA4b in mice inhibited nNOS activity and reduced cardiac hypertrophy by inhibiting calcineurin. Here we examine temporally regulated cardiac-specific overexpression of
Hui-Hsing Hung et al.
Molecular and cellular neurosciences, 82, 35-45 (2017-04-22)
Zinc ion (Zn2+), the second most abundant transition metal after iron in the body, is essential for neuronal activity and also induces toxicity if the concentration is abnormally high. Our previous results show that exposure of cultured cortical neurons to
Potent and selective inhibition of neuronal nitric oxide synthase by N omega-propyl-L-arginine.
H Q Zhang et al.
Journal of medicinal chemistry, 40(24), 3869-3870 (1997-12-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service