Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2025

Sigma-Aldrich

AH-7614

≥98% (HPLC)

Synonym(s):

4-Methyl-N-9H-xanthen-9-yl-benzenesulfonamide, AH 7614, Compound 39, NSC 31171, p-Toluenesulfonamide, N-xanthen-9-yl-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17NO3S
CAS Number:
Molecular Weight:
351.42
MDL number:
UNSPSC Code:
41121800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(NC1C2=C(C=CC=C2)OC3=C1C=CC=C3)(C4=CC=C(C)C=C4)=O

InChI

1S/C20H17NO3S/c1-14-10-12-15(13-11-14)25(22,23)21-20-16-6-2-4-8-18(16)24-19-9-5-3-7-17(19)20/h2-13,20-21H,1H3

InChI key

OZCQEUZTOAAWDK-UHFFFAOYSA-N

Biochem/physiol Actions

AH-7614 is a potent negative allosteric modulator (NAM) of the long-chain free fatty acid receptor FFA4 that prevents differentiation of a mouse mesenchymal stem cell line toward an adipocyte phenotype. AH-7614 (Compound 39) was first described as antagonist of FFA4 that blocks activation of FFA4 by polyunsaturated, ω-6 fatty acid, linoleic acid and GSK137647A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven M Sparks et al.
Bioorganic & medicinal chemistry letters, 24(14), 3100-3103 (2014-06-03)
The exploration of a diarylsulfonamide series of free fatty acid receptor 4 (FFA4/GPR120) agonists is described. This work led to the identification of selective FFA4 agonist 8 (GSK137647A) and selective FFA4 antagonist 39. The in vitro profile of compounds 8
Kenneth R Watterson et al.
Molecular pharmacology, 91(6), 630-641 (2017-04-08)
High-affinity and selective antagonists that are able to block the actions of both endogenous and synthetic agonists of G protein-coupled receptors are integral to analysis of receptor function and to support suggestions of therapeutic potential. Although there is great interest
S Villegas-Comonfort et al.
Prostaglandins, leukotrienes, and essential fatty acids, 117, 1-10 (2017-02-27)
Arachidonic acid increased intracellular calcium, in cells expressing green fluorescent protein-tagged human FFA4 receptors, with an EC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service