Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2015

Sigma-Aldrich

MM11253

≥98% (HPLC)

Synonym(s):

2-(2-Carboxy-6-naphthalenyl)-2-(5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphthalen-2-yl)-1,3-dithiolane, 6-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1,3-dithiolan-2-yl]-2-naphthalenecarboxylic acid, MM 11253, MM-11253, SR 11253, SR11253

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30O2S2
CAS Number:
Molecular Weight:
462.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C1=CC(C=CC(C2(SCCS2)C3=CC(C(C)(C)CCC4(C)C)=C4C=C3)=C5)=C5C=C1)=O

InChI

1S/C28H30O2S2/c1-26(2)11-12-27(3,4)24-17-22(9-10-23(24)26)28(31-13-14-32-28)21-8-7-18-15-20(25(29)30)6-5-19(18)16-21/h5-10,15-17H,11-14H2,1-4H3,(H,29,30)

InChI key

DPVGXUNWCVEIGT-UHFFFAOYSA-N

Biochem/physiol Actions

MM11253 (SR11253) is a high-affinity RARγ-selective retinoid acid (RA) receptor antagonist (IC50 = 44 nM against ATRA for binding RARγ; IC50 =1 μM in case of RARα, RARβ, RXRα) that prevents >90% growth inhibitory effect of retinoids AHPN/CD437 (1 μM in CA-OV3 & SK-SKOV3 cultures) and M11389 (1 μM in SCC-25 squamous cell carcinoma cultures) when used at equal concentration. MM11253 also effectively blocks the inhibitory effect of R667 (RARγ agonist) against IL-1β-induced MMP production from rabbit corneal fibroblasts (80% vs. 11% inhibition by 0,1 nM R667, respectively, without or with 0.03 nM MM11253).
RARγ selective antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service