Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2665

Sigma-Aldrich

AGN194310

≥98% (HPLC)

Synonym(s):

4-[2-[4-(4-Ethylphenyl)-2,2-dimethyl-2H-1-benzothiopyran-6-yl]ethynyl]benzoic acid, 4-[[4-(4-Ethylphenyl)-2,2-dimethyl-2H-1-benzothiopyran-6-yl]ethynyl]benzoic acid, AGN 194310, AGN-194310, IRX4310, VTP 194310

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H24O2S
CAS Number:
Molecular Weight:
424.55
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC=C(C#CC2=CC3=C(SC(C)(C=C3C4=CC=C(CC)C=C4)C)C=C2)C=C1

InChI

1S/C28H24O2S/c1-4-19-7-12-22(13-8-19)25-18-28(2,3)31-26-16-11-21(17-24(25)26)6-5-20-9-14-23(15-10-20)27(29)30/h7-18H,4H2,1-3H3,(H,29,30)

InChI key

LHUPKWKWYWOMSK-UHFFFAOYSA-N

Biochem/physiol Actions

AGN194310 (IRX4310; VTP 194310) is a retinoic acid receptor (RAR) antagonist that targets all three RAR subtypes with higher affinity (RARα/β/γ Kd = 3/2/5 nM) than all-trans retinoic acid/ATRA (RARα/β/γ Kd = 15/13/18 nM) without affecting RXR subtypes. AGN194310 effectively blocks RAR agonist TTNPB-induced transcription activity (using RARα, RARβ, or RARγ transfected cells) and prevents TTNPB toxicity in guinea pigs in vivo (by 79/84/94% at AGN194310/TTNPB ratios of 0.5/2/8). AGN194310 potentiates differentiation in osteoblast cultures (1 μM) and daily oral administration (0.5 mg/kg/d for a10-d period) is reported to cause impaired radial bone growth among 8-wk-old mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service