Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2013

Sigma-Aldrich

RU 24969 hemisuccinate

≥98% (HPLC)

Synonym(s):

1H-Indole, 5-methoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-, butanedioate (2:1), 5-Methoxy-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-Indole hemisuccinate, 5-Methoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O · 0.5C4H6O4
CAS Number:
Molecular Weight:
287.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to yellow-orange

solubility

H2O: 2 mg/mL, clear (warmed)

shipped in

ambient

storage temp.

room temp

SMILES string

COC1=CC2=C(NC=C2C3=CCNCC3)C=C1.OC(CCC(O)=O)=O.COC4=CC5=C(NC=C5C6=CCNCC6)C=C4

InChI

1S/2C14H16N2O.C4H6O4/c2*1-17-11-2-3-14-12(8-11)13(9-16-14)10-4-6-15-7-5-10;5-3(6)1-2-4(7)8/h2*2-4,8-9,15-16H,5-7H2,1H3;1-2H2,(H,5,6)(H,7,8)

InChI key

BNYJLRQAWCQJOM-UHFFFAOYSA-N

Biochem/physiol Actions

RU-24969 is a potent and selective agonist at the 5-HT1A and 5-HT1B receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily V Ho et al.
Psychopharmacology, 233(1), 57-70 (2015-10-02)
Serotonin-1B receptor (5-HT1BR) agonist treatment induces obsessive-compulsive disorder (OCD)-like behaviors including locomotor stereotypy, prepulse inhibition deficits, and delayed alternation disruptions, which are selectively prevented by clinically effective OCD treatment. However, the role of 5-HT1BRs in modulating other repetitive behaviors or
Dane Aronsen et al.
Pharmacology, biochemistry, and behavior, 124, 1-4 (2014-05-23)
RU 24969 is a widely used, but non-selective, 5-HT1B/1A agonist that decreases fluid consumption and increases forward locomotion. The mechanism underlying these behavioural responses is not, however, well understood. Accordingly, effects of the selective 5-HT1A and 5-HT1B antagonists, WAY 100635
Dane Aronsen et al.
Psychopharmacology, 233(8), 1339-1347 (2016-02-10)
3,4 Methylenedioxymethamphetamine (MDMA) preferentially stimulates the release of serotonin (5-HT) that subsequently produces behavioral responses by activation of post-synaptic receptor mechanisms. The 5-HT1A and 5-HT1B receptors are both well localized to regulate dopamine (DA) release, and have been implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service