Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1990

Sigma-Aldrich

GW806742X

≥98% (HPLC)

Synonym(s):

3-(4-(Methyl(4-(3-(4-(trifluoromethoxy)phenyl)ureido)phenyl)amino)pyrimidin-2-ylamino)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22F3N7O4S
CAS Number:
Molecular Weight:
573.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=NC(NC2=CC(S(=O)(N)=O)=CC=C2)=NC=C1)C3=CC=C(NC(NC4=CC=C(OC(F)(F)F)C=C4)=O)C=C3

InChI

1S/C25H22F3N7O4S/c1-35(22-13-14-30-23(34-22)31-18-3-2-4-21(15-18)40(29,37)38)19-9-5-16(6-10-19)32-24(36)33-17-7-11-20(12-8-17)39-25(26,27)28/h2-15H,1H3,(H2,29,37,38)(H,30,31,34)(H2,32,33,36)

InChI key

SNRUTMWCDZHKKM-UHFFFAOYSA-N

Biochem/physiol Actions

ATP site-targeting kinase inhibitor and an anti-necroptosis agent that targets MLKL pseudokinase domain.
Originally characterized an ATP site-targeing kinase inhibitor (IC50 in nM = 2/VEGFR2, 15/SRC, 47/C-FMS, 851/GSK3, 9016/ERBB2, 9120/FGFR) with potent antiproliferation activity against cancer cultures (IC50 in nM = 5/HUVEC-v, 424/HUVEC-b, 81/HEF, 453/MDA468, 470/A375P, 693/HT29, 734/PC3), GW806742X is now better known as Compound 1 for its anti-necroptosis activity via affinity interaction with MLKL pseudokinase domain (Kd = 9.3 μM), thereby preventing the 4HB domain from exerting its necroptototic effect following RIPK3-mediated pseudokinase domain phosphorylation. GW806742X inhibits TSQ (1 ng/mL TNF, 500 nM Smac-mimetic, 10 μM Q-VD-Oph) treatment-induced necroptosis of mouse dermal fibroblasts (IC50 <50 nM) with >50-fold greater potency than Nec-1, while reduced efficiency is only observed in the presence of supraphysiological TNF concentrations of 100 ng/mL (IC50 = 100-500 nM; Max efficacy ~50%).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service