Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1532

Sigma-Aldrich

Cinaciguat hydrochloride

≥98% (HPLC)

Synonym(s):

4-[((4-Carboxybutyl){2-[(4-phenethylbenzyl)oxy]phenethyl}amino)methyl[benzoic]acid hydrochloride, 4-[[(4-Carboxybutyl)[2-[2-[[4-(2-phenylethyl)phenyl]methoxy]phenyl]ethyl]amino]methyl]-benzoic acid hydrochloride, BAY 58-2667 hydrochloride, BAY582667 hydrochloride, UNII-59K0Y58UAD hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H39NO5 · HCl
CAS Number:
Molecular Weight:
602.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

OC(C(C=C1)=CC=C1CN(CCCCC(O)=O)CCC2=CC=CC=C2OCC3=CC=C(CCC4=CC=CC=C4)C=C3)=O.[H]Cl

InChI

1S/C36H39NO5.ClH/c38-35(39)12-6-7-24-37(26-30-19-21-33(22-20-30)36(40)41)25-23-32-10-4-5-11-34(32)42-27-31-17-15-29(16-18-31)14-13-28-8-2-1-3-9-28;/h1-5,8-11,15-22H,6-7,12-14,23-27H2,(H,38,39)(H,40,41);1H

InChI key

LLHMBJOVHATVSP-UHFFFAOYSA-N

Biochem/physiol Actions

Cinaciguat activates sGC (soluble guanylate cyclase) independently of NO. Cinaciguat binds to sGC′s NO-sensory H-NOX domain when it is heme-depleted. Also, the compound Cinaciguat protects sGC from oxidation-induced ubiquitin-triggered degradation.
Cinaciguat, also known as BAY 58-2667, is a novel activator of guanylate cyclase. It is used in the treatment of acute decompensated heart failure (ADHF). It has been reported that cinaciguat possesses cardiovascular effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang Mueck et al.
Clinical pharmacokinetics, 49(2), 119-129 (2010-01-14)
Cinaciguat (BAY 58-2667) is a novel soluble guanylate cyclase activator in clinical development for the treatment of acute decompensated heart failure (ADHF). In patients with ADHF, intravenously administered cinaciguat results in potent unloading of the heart with arterial vasodilation. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service