Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1518

Sigma-Aldrich

SID 3712249

≥95% (HPLC)

Synonym(s):

1,8-Diamino-3,6-di-1-pyrrolidinyl-2,7-naphthyridine-4-carbonitrile, MLS000054131, MiR-544 Inhibitor 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N7
CAS Number:
Molecular Weight:
323.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C2C(C=C(N3CCCC3)N=C2N)=C(C#N)C(N4CCCC4)=N1

InChI

1S/C17H21N7/c18-10-12-11-9-13(23-5-1-2-6-23)21-15(19)14(11)16(20)22-17(12)24-7-3-4-8-24/h9H,1-8H2,(H2,19,21)(H2,20,22)

InChI key

SSWVPYMSHHEYAB-UHFFFAOYSA-N

Biochem/physiol Actions

SID 3712249 (MiR-544 Inhibitor 1) is an inhibitor of the biogenesis of microRNA-544 (miR-544). MiR-544 represses expression of mTOR, promoting tumor cell survival in a hypoxic environment. Inhibition of miR-544 processing with MiR-544 Inhibitor 1 caused apoptosis in triple negative breast cancer cells in response to hypoxic stress, sensitized their response to 5-fluorouracil, and blocked their growth after transplantion into immunodeficient mice. SID 3712249 (MiR-544 Inhibitor 1) is believed to bind directly to the precursor miRNA, blocking production of the mature microRNA and resulting in decreased miR-544, HIF-1α, and ATM transcripts. MiR-544 Inhibitor 1 was as selective and 25-fold more potent than a 2′-O-methyl RNA antagomir.

Other Notes

light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service