Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510440

Sigma-Aldrich

4-Cyanobenzophenone

97%

Synonym(s):

4-Benzoylbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4OCC6H5
CAS Number:
Molecular Weight:
207.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

110-114 °C (lit.)

functional group

ketone
nitrile
phenyl

SMILES string

O=C(c1ccccc1)c2ccc(cc2)C#N

InChI

1S/C14H9NO/c15-10-11-6-8-13(9-7-11)14(16)12-4-2-1-3-5-12/h1-9H

InChI key

YSZWJJANSNFQMM-UHFFFAOYSA-N

General description

4-Cyanobenzophenone is a benzophenone derivative that can be prepared by treating 4-cyanobenzoyl chloride with benzene. Its reduction under electrochemical condition led to the formation of 4-t-butyl-benzophenone and 1-(4-cyanophenyl)-2,2-dimthyl-1-phenylpropan-1-ol. Its efficiency as triplet sensitizer for norbornadiene(N)→quadriciane(Q) and Q→N photoisomerization has been assessed.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemically induced aromatic nucleophilic substitution.
Pinson J and Saveant JM.
Journal of the American Chemical Society, 100(5), 1506-1510 (1978)
Mechanistic approach to the sensitization process of aromatic ketones in the isomerization between norbornadiene and quadricyclane.
Arai T, et al.
Bulletin of the Chemical Society of Japan, 60(8), 2937-2943 (1987)
Electrochemical t-Butylation of Some Aromatic Ketones.
Kristensen LH and Lund H.
Acta Chemica Scandinavica. Series B, 33, 735-741 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service