Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1471

Sigma-Aldrich

SN50 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

AAVALLPAVLLALLAPVQRKRQKLMP trifluoroacetate salt, H-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Gln-Arg-Lys-Arg-Gln-Lys-Leu-Met-Pro-OH trifluoroacetate salt, NF-KB SN50 cell permeable inhibitory peptide trifluoroacetate salt, Nuclear Factor NF-KB Inhibitor SN50 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C129H230N36O29S · xC2HF3O2
CAS Number:
Molecular Weight:
2781.50 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

film

color

white

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C129H230N36O29S/c1-64(2)57-87(156-117(183)92(62-69(11)12)158-123(189)100(72(17)18)161-106(172)78(25)143-119(185)94-41-35-54-164(94)126(192)93(63-70(13)14)159-118(184)90(60-67(7)8)154-104(170)76(23)144-121(187)99(71(15)16)160-105(171)77(24)141-102(168)74(21)132)113(179)142-75(22)103(169)153-89(59-66(5)6)116(182)157-88(58-65(3)4)114(180)145-79(26)124(190)163-53-34-42-95(163)120(186)162-101(73(19)20)122(188)151-85(45-47-98(134)167)112(178)149-82(39-32-51-139-128(135)136)108(174)146-80(37-28-30-49-130)107(173)148-83(40-33-52-140-129(137)138)109(175)150-84(44-46-97(133)166)111(177)147-81(38-29-31-50-131)110(176)155-91(61-68(9)10)115(181)152-86(48-56-195-27)125(191)165-55-36-43-96(165)127(193)194/h64-96,99-101H,28-63,130-132H2,1-27H3,(H2,133,166)(H2,134,167)(H,141,168)(H,142,179)(H,143,185)(H,144,187)(H,145,180)(H,146,174)(H,147,177)(H,148,173)(H,149,178)(H,150,175)(H,151,188)(H,152,181)(H,153,169)(H,154,170)(H,155,176)(H,156,183)(H,157,182)(H,158,189)(H,159,184)(H,160,171)(H,161,172)(H,162,186)(H,193,194)(H4,135,136,139)(H4,137,138,140)/t74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,99-,100-,101-/m0/s1

InChI key

FAWLNURBQMTKEB-URDPEVQOSA-N

Application

SN50 trifluoroacetate salt has been used as a nuclear factor-kappa B (NF-κB) inhibitor:
  • to study its effects on the reactive oxygen species generation in human keratinocytes
  • to analyze its effects on mRNA expression in the receptor for advanced glycation end products (RAGE) in hCMEC/D3 blood-brain barrier (BBB) cell line
  • to study its effects on macrophage polarization

Biochem/physiol Actions

SN50 is an NF-κB cell permeable inhibitory peptide. The SN50 sequence contains the nuclear localization sequence (NLS residues 360-369) of the transcription factor NFκB p50 linked to a peptide cell-permeabilization sequence, the hydrophobic region (h-region) of the signal peptide of Kaposi fibroblast growth factor (K-FGF). SN50 Peptide inhibits translocation of the NF-κB active complex into the nucleus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Fang Nie et al.
Journal of immunology research, 2018, 6285813-6285813 (2018-10-03)
This study investigated the alterations in macrophage polarization in patients with endometriosis as well as the underlying molecular mechanisms. Peritoneal washings, serum samples, and endometrial tissues were collected from endometriosis patients and control subjects. Endometrial stromal cells (ESCs) were isolated
Jinjin Dong et al.
Oncotarget, 8(65), 109161-109174 (2018-01-10)
Vitiligo is an acquired depigmentary skin inflammatory disorder. The pathogenesis of inflammatory skin disease involves the release of cytokines from keratinocytes, including interleukin (IL)-1β. IL-22 belongs to a family of cytokines structurally related to IL-10, including IL-19, IL-20, IL-24, and
Fan Zeng et al.
Journal of neurochemistry, 158(3), 724-736 (2020-05-23)
Cerebrovascular-related amyloidogenesis is found in over 80% of Alzheimer's disease (AD) cases, and amyloid β (Aβ) generation is increased in the peripheral macrophages during infection of Porphyromonas gingivalis (P. gingivalis), a causal bacterium for periodontitis. In this study, we focused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service