Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1138

Sigma-Aldrich

IKK-16 dihydrochloride

≥98% (HPLC)

Synonym(s):

N-(4-Pyrrolidin-1-yl-piperidin-1-yl)-[4-(4-benzo[b]thiophen-2-yl-pyrimidin-2-ylamino)phenyl]carboxamide dihydrochloride, IKK16 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29N5OS · 2HCl
CAS Number:
Molecular Weight:
556.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCC(N2CCCC2)CC1)C3=CC=C(NC4=NC=CC(C5=CC(C=CC=C6)=C6S5)=N4)C=C3.C

InChI

1S/C28H29N5OS.CH4/c34-27(33-17-12-23(13-18-33)32-15-3-4-16-32)20-7-9-22(10-8-20)30-28-29-14-11-24(31-28)26-19-21-5-1-2-6-25(21)35-26;/h1-2,5-11,14,19,23H,3-4,12-13,15-18H2,(H,29,30,31);1H4

InChI key

YJGZKHHCMOHIFA-UHFFFAOYSA-N

Application

IKK-16 dihydrochloride has been used as an nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor kinase inhibitor (IκB) in B220+ CKO or CreWT naive B cells. It has also been used as a nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB inhibitor) in poly(I:C) transfection-induced retinoic acid-inducible gene (RIG-I), melanoma differentiation-associated gene 5 (MDA-5), and caspase-11 expressing mouse embryonic fibroblasts (MEFs).

Biochem/physiol Actions

IKK-16 is a selective inhibitor of IKB kinase (IKK), part of the upstream NF-KB signal transduction cascade involved in the cellular response to inflammation. IKK-16 has IC50 values of 40, 70 and 200 nM for IKKβ (IKK-2), IKK complex and IKKα (IKK-1) respectively. In a mouse sepsis model, IKK-16 reduced the multiple organ dysfunction, showing an antiinflammatory and endothelial protective effect likely mediated by inhibition of inflammation through NF-KB. IKK-16 has also been found to have some activity as an inhibitor of protein kinase D (PKD) and of multidrug resistance protein 1 (MDR1, ABCB1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenyi Su et al.
Cell death and differentiation, 26(6), 1077-1088 (2018-10-21)
ABIN-1 (encoded by the gene Tnip1) is a ubiquitin-binding protein that can interact with ubiquitin-editing enzyme A20 (encoded by the gene TNFAIP3) to restrain the activation of necroptosis and NF-κB activation. Genetic variants in the genes Tnip1 and TNFAIP3 are
Robert R Haines et al.
Journal of immunology (Baltimore, Md. : 1950), 203(7), 1867-1881 (2019-09-08)
Marginal zone B cells (MZB) are a mature B cell subset that rapidly respond to blood-borne pathogens. Although the transcriptional changes that occur throughout MZB development are known, the corresponding epigenetic changes and epigenetic modifying proteins that facilitate these changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service