Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1050

Sigma-Aldrich

DC260126

≥98% (HPLC)

Synonym(s):

N-(4-Butylphenyl)-4-fluoro-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18FNO2S
CAS Number:
Molecular Weight:
307.38
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)[S](=O)(=O)Nc2ccc(cc2)CCCC

InChI

1S/C16H18FNO2S/c1-2-3-4-13-5-9-15(10-6-13)18-21(19,20)16-11-7-14(17)8-12-16/h5-12,18H,2-4H2,1H3

InChI key

CNGHPXKWPGIDSK-UHFFFAOYSA-N

Biochem/physiol Actions

DC260126 is a FFA1 (GPR40) selective antagonist.
DC260126 is a FFA1 (GPR40) selective antagonist. In FFA1-expressing CHO cells, DC260126 blocks linoleic acid-induced Ca2+ elevation and ERK phosphorylation (IC50 = 6028 μM), as well as palmitic acid potentiated glucose-stimulated insulin secretion. The compound DC260126 blocks MIN6 β cells from palmitic acid-induced ER stress and apoptosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Zhao et al.
Microbiome, 6(1), 107-107 (2018-06-16)
The gut microbiota is closely associated with gastrointestinal (GI) motility disorder, but the mechanism(s) by which bacteria interact with and affect host GI motility remains unclear. In this study, through using metabolomic and metagenomic analyses, an animal model of neonatal

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service