Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML0354

Sigma-Aldrich

Neuronostatin trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

L-Leucyl-L-arginyl-L-glutaminyl-L-phenylalanyl-L-leucyl-L-glutaminyl-L-lysyl-L-seryl-L-leucyl-L-alanyl-L-alanyl-L-alanyl-L-alaninamide trifluoroacetate salt, Leu-Arg-Gln-Phe-Leu-Gln-Lys-Ser-Leu-Ala-Ala-Ala-Ala-NH2 trifluoroacetate salt, Neuronostatin-13 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H110N20O16 · xC2HF3O2
CAS Number:
Molecular Weight:
1415.68 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

color

white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Neuronostatin is a peptide hormone encoded by somatostatin gene that is involved in regulation of neuronal function, blood pressure, food intake and drinking behavior. Data indicates that neuronostatin regulates cardiac contractile function and cardiomyocyte survival. Also neuronostatin significantly inhibited both food and water intake when administered centrally in adult male rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Somatostatin (somatotropin release-inhibiting factor, SRIF) is a cyclic peptide widely expressed throughout the CNS, endocrine tissues, and gastrointestinal tract.

Somatostatin (somatotropin release-inhibiting factor, SRIF) is a cyclic peptide widely expressed throughout the CNS, endocrine tissues, and gastrointestinal tract.

Somatostatin (somatotropin release-inhibiting factor, SRIF) is a cyclic peptide widely expressed throughout the CNS, endocrine tissues, and gastrointestinal tract.

Somatostatin (somatotropin release-inhibiting factor, SRIF) is a cyclic peptide widely expressed throughout the CNS, endocrine tissues, and gastrointestinal tract.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service