Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01044

Sigma-Aldrich

Nevadensin

≥90% (LC/MS-ELSD)

Synonym(s):

5,7-dihydroxy-6,8-dimethoxy-2-(4-methoxyphenyl)chromen-4-one, Ethoxychelerythrine, Lysionotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

344.32

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c([c](cc1c3ccc(cc3)OC)=O)c(c(c(c2OC)O)OC)O

InChI

1S/C18H16O7/c1-22-10-6-4-9(5-7-10)12-8-11(19)13-14(20)17(23-2)15(21)18(24-3)16(13)25-12/h4-8,20-21H,1-3H3

InChI key

KRFBMPVGAYGGJE-UHFFFAOYSA-N

General description

Nevadensin is a hydroxyflavone-flavonoid compound commonly found in plants such as Ocimum Sp., Lysionotus pauceflora, Hyptis albida, Rosa centifolia, Calanticaria bicolor, and Gardenia resinifera. Existing research suggests that this plant-derived metabolite exerts various biological activities, including antibacterial, anti-estrogenic, and anti-infective properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Nevadensin exhibits a wide range of significant biological activities including hypotensive, anti-tubercular, antimicrobial, anti-inflammatory, anti-tumour and anti-cancer activities. It displays high inhibition potency and selectivity towards hCE1.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service