Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB01010

Sigma-Aldrich

Marrubiin

≥90% (LC/MS-ELSD)

Synonym(s):

(1R,4S,8S,9R,10R,12R)-9-[2-(furan-3-yl)ethyl]-9-hydroxy-4,8,10-trimethyl-2-oxatricyclo[6.3.1.04,12]dodecan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

332.43

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C20H28O4/c1-13-11-15-16-18(2,17(21)24-15)7-4-8-19(16,3)20(13,22)9-5-14-6-10-23-12-14/h6,10,12-13,15-16,22H,4-5,7-9,11H2,1-3H3/t13-,15-,16+,18+,19+,20-/m1/s1

InChI key

HQLLRHCTVDVUJB-OBHOOXMTSA-N

General description

Marrubiin is a labdanoid lactone commonly derived from Marrubium Sp. plants, including M. globosum, M. alysson, M. vulgare, and M. anisodon. Existing research suggests that this plant-derived metabolite possesses various biological properties, including antioxidant, antifungal, gastroprotective, immunomodulatory, cardioprotective, anti-diabetic, and analgesic activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Based on existing research, Marrubiin has demonstrated the ability to inhibit the production of pro-inflammatory cytokines and reduce the activity of enzymes involved in oxidative stress. Additionally, it has been observed to lower blood pressure and provide protection against ischemic heart injury.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Meyre-Silva et al.
Farmaco (Societa chimica italiana : 1989), 60(4), 321-326 (2005-04-26)
Marrubiin, a furane labdane diterpene, is the main analgesic compound present in Marrubium vulgare, a medicinal plant used in Brazil and other countries to treat several ailments. Considering its important pharmacological action, as well as its high yield, some structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service