Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00086

Sigma-Aldrich

Columbin

≥95% (LC/MS-ELSD)

Synonym(s):

Isocolumbin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@]12C[C@H](OC(=O)[C@@H]1CC[C@]3(C)[C@H]2[C@@H]4OC(=O)[C@@]3(O)C=C4)c5ccoc5

InChI

1S/C20H22O6/c1-18-9-14(11-5-8-24-10-11)25-16(21)12(18)3-6-19(2)15(18)13-4-7-20(19,23)17(22)26-13/h4-5,7-8,10,12-15,23H,3,6,9H2,1-2H3/t12-,13+,14-,15-,18+,19+,20-/m0/s1

InChI key

AALLCALQGXXWNA-QJNFORGASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siddig Ibrahim Abdelwahab et al.
European journal of pharmacology, 678(1-3), 61-70 (2012-01-10)
Columbin, a diterpenoid furanolactone, was isolated purely for the first time from the plant species Tinspora bakis. The anti-inflammatory effects of columbin were studied in vitro, in silico and in vivo. The effect of columbin on nitric oxide was examined
Qirong Shi et al.
Biomedical chromatography : BMC, 21(6), 642-648 (2007-03-09)
Columbin is an important component isolated from Radix Tinosporae. It has been demonstrated to possess many pharmacological activities, including anti-inflammation, antitumor and inhibition of enzyme activity in vivo. The purpose of the present study was to examine in vivo pharmacokinetics
Hiroyuki Kohno et al.
Cancer letters, 183(2), 131-139 (2002-06-18)
The modifying effect of dietary administration of a diterpenoid furanolactone columbin isolated from the crude drug Calumbae Radix (the root of Jateorhiza columba MIERS, Menispermacea) on azoxymethane (AOM)-induced was investigated in male F344 rats. Animals were initiated with AOM (three
K Wada et al.
Biological & pharmaceutical bulletin, 18(4), 634-636 (1995-04-01)
In a screening series of bioactive components in edible and medicinal plants, we found that Calumbae Radix (Colombo root, 1% powdered feed, for 5 d) and its component, columbin (20-40 mg/kg/d, for 5 d, orally), shortened the sleeping time induced
A J Nok et al.
Journal of enzyme inhibition and medicinal chemistry, 17(1), 55-59 (2002-10-09)
The inhibitory effects of a naturally occurring diterpenoid furanolactone, columbin, on partially purified acidic phospholipase A2 (PLA2) from Naja nigricolis was investigated. Columbin inhibited the N. nigricolis PLA2 in a dose related pattern with an IC50 value of 2.5 microM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service