Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00209

Sigma-Aldrich

Nimbidiol

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O3
CAS Number:
Molecular Weight:
274.35
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)CCC[C@@]2(C)C1CC(=O)c3cc(O)c(O)cc23

InChI

1S/C17H22O3/c1-16(2)5-4-6-17(3)11-8-14(20)13(19)7-10(11)12(18)9-15(16)17/h7-8,15,19-20H,4-6,9H2,1-3H3/t15?,17-/m1/s1

InChI key

JMBKXUYCBVKSSY-OMOCHNIRSA-N

General description

Natural product derived from plant source.
Nimbidiol is a modified diterpenoid. It is isolated from the root-bark of Azadirachta indica (Indian ‘neem′).

Biochem/physiol Actions

Nimbidiol has an ability to inhibit intestinal glucosidases, including maltase-glucoamylase, sucrase-isomaltase, lactase, trehalase and fungal α-glucosidases. Thus, this bioactive molecule has all the potential to be used as an anti-diabetic drug

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nimbidiol, a modified diterpenoid of the root-bark of Azadirachta indica
Majumder P L, et al.
Phytochemistry, 26(11), 3021-3023 (1987)
Abhishek Mukherjee et al.
Journal of enzyme inhibition and medicinal chemistry, 28(5), 900-910 (2012-07-19)
Azadirachta indica, used in antidiabetic herbal drugs, was reported to contain α-glucosidase inhibitor. Bioassay guided purification characterized the inhibitor as nimbidiol (a diterpenoid), present in root and stem-bark of the tree. Nimbidiol inhibited intestinal (mammalian) maltase-glucoamylase, sucrase-isomaltase, lactase, trehalase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service