Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01053

Sigma-Aldrich

Arnicolide C

≥90% (LC/MS-ELSD)

Synonym(s):

11,13-Dihydrohelenalin isobutyrate, 6-O-Isobutyryl-11,13-dihydrohelenalin, Isobutyroylplenolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O5
CAS Number:
Molecular Weight:
334.41
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

334.41

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Arnicolide C, also known as Isobutyroylplenolin, is a sesquiterpene lactone commonly found in plants such as Centipeda minima and Arnica montana. Current research suggests that this naturally occurring metabolite may possess various biological activities, including anti-inflammatory, antiallergy, antitumor, anticancer, and antiprotozoal properties.

Application

Arnicolide C is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biologically active constituents of Centipeda minima: isolation of a new plenolin ester and the antiallergy activity of sesquiterpene lactones.
J B Wu et al.
Chemical & pharmaceutical bulletin, 33(9), 4091-4094 (1985-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service