Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00081

Sigma-Aldrich

Wogonoside

≥95% (LC/MS-ELSD)

Synonym(s):

5,7-Dihydroxy 8-methoxyflavone 7-glucuronide, Glychionide B, Oroxindin, Wogonin 7-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O11
CAS Number:
Molecular Weight:
460.39
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1c(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(O)=O)cc(O)c3C(=O)C=C(Oc13)c4ccccc4

InChI

1S/C22H20O11/c1-30-18-13(32-22-17(27)15(25)16(26)20(33-22)21(28)29)8-11(24)14-10(23)7-12(31-19(14)18)9-5-3-2-4-6-9/h2-8,15-17,20,22,24-27H,1H3,(H,28,29)/t15-,16-,17+,20-,22+/m0/s1

InChI key

LNOHXHDWGCMVCO-NTKSAMNMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

Wogonoside, a flavanoid, is known for its anti-inflammatory properties. Wogonoside may have therapeutic potential for use in diseases which involve the development of inflammation and angiogenesis through its inhibition of LPS-induced angiogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Yan et al.
Chinese medicine, 13, 54-54 (2018-11-06)
Microbial fermentation has been widely applied in traditional Chinese medicine (TCM) for thousands of years in China. Various beneficial effects of fermentation for applications in TCM or herbals have been reported, such as enhanced anti-ovarian cancer, antioxidative activity, and neuroprotective
Zhenyu Zhu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(24), 2184-2190 (2010-07-21)
The aim of this study was to compare the pharmacokinetics of baicalin and wogonoside in rats following oral administration of Xiaochaihu Tang (Minor Radix Bupleuri Decoction) and Radix scutellariae extract. Thus, a specific LC-MS method was developed and validated for
R Xiao et al.
Human & experimental toxicology, 36(11), 1169-1176 (2016-12-13)
Wogonoside is the main flavonoid of the traditional Chinese medicinal herb Scutellaria baicalensis Georgi and has been found to induce growth suppression in myelogenous leukemia cells. However, its activity in T acute lymphoblastic leukemia (T-ALL) is still unclear. In this
Xiao-Yan Wei et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1061-1062, 248-255 (2017-08-02)
A rapid, sensitive, and selective UPLC-TQ-MS/MS method was established and validated for the determination and pharmacokinetic investigation of rhein, coptisine, berberine, palmatine, baicalin and wogonoside from Sanhuang Xiexin Decoction (SXD) extracts. A Waters BEH C
Ying Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(20-21), 1820-1826 (2009-06-06)
A specific ultra-performance liquid chromatography-mass spectrometry (UPLC-MS) method was developed for the simultaneous determination of puerarin, daidzein, baicalin, wogonoside and liquiritin in rat plasma. Chromatographic separation was performed on a C(18) column packed with 1.7 microm particles by a linear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service