Skip to Content
MilliporeSigma

Skip To

SHM04

MISSION® ExpressMag® Super Magnetic Plate

Increases transduction efficiency

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.51
UNSPSC Code:
41106609
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

MISSION®

storage temp.

room temp

General description

To view protocols from Sigma collaborators for ExpressMag system visit the link below.
www.sigmaaldrich.com/mission-protocols

Application

MISSION® ExpressMag® Super Magnetic Plate exerts a magnetic force used to attract magnetic nanoparticles to the surface of cells in culture.
MISSION® ExpressMag® Super Magnetic Plate exerts a magnetic force used to attract magnetic nanoparticles to the surface of cells in culture. This product can be used in conjunction with standard sized well plates, T-25 through T-75 tissue culture flasks, and up to 5 cm dishes.
Magnets should be kept away from magnetic storage devices, such as hard disks, ID cards, and credit cards, and from electronic devices and ferromagnetic materials. Persons with cardiac pacemakers should not work with these magnets.
It has been used to culture rat neural stem cells for transfection of green fluorescent protein.

Legal Information

ExpressMag is a registered trademark of Sigma-Aldrich Co. LLC
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Do not autoclave! Clean surface with 70% isopropyl alcohol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SHM05SHM0328878U
description

Increases transduction efficiency

description

Increases transduction efficiency

description

Increases transduction efficiency

description

carrier gas kit (includes SU861026 + SU861011)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

-

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. What is the typical fold increase in transduction efficiency when using ExpressMag beads?

    1 answer
    1. The transduction efficiency increase observed when using the ExpressMag products will depend on the cell line. It is reasonable to expect a roughly 10-fold or greater increase.

      Helpful?

  2. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What MISSION products have been successfully introduced into cells using ExpressMag® beads?

    1 answer
    1. MISSION Lentiviral Particles (various targets/clones), SHC002V, SHC003V and SHC008V have been successfully introduced into cells using ExpressMag beads.

      Helpful?

  5. What other viruses have been successfully introduced into cells via the use of ExpressMag beads?

    1 answer
    1. Adenovirus, SIV, Retrovirus, Measles virus and HIV have all been successfully introduced into cells using ExpressMag.

      Helpful?

  6. What cell lines have been transduced successfully using ExpressMag Beads?

    1 answer
    1. The following cell types have been successfully transduced using ExpressMag Beads: PBL, PBMC, T Cells, GMB primary tumor cells, A549, NYGM, T98G, U251, U87,YH-13, YK6-1, KS-1, NIH3T3, K562, Vero, HeLa, CHO, L929, B95a, H9, HUVEC, CD4+ Primary Human T-Cells, Human Bone Marrow Macrophages, Primary Keratinocytes, SKOV3, MM-AN, MEF and CD 34+.

      Helpful?

Reviews

No rating value

Active Filters