Skip to Content
MilliporeSigma

SHM05

MISSION® ExpressMag® 96-Well Magnet

Increases transduction efficiency

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41106609
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

MISSION®

storage temp.

room temp

General description

To view protocols from Sigma collaborators for ExpressMag system visit the link below.
www.sigmaaldrich.com/mission-protocols

Application

MISSION® ExpressMag® 96-Well Magnet exerts a magnetic force used to attract magnetic nanoparticles to the surface of cells in culture. This product is optimized for use with 96-well tissue culture plates.

Legal Information

ExpressMag is a registered trademark of Sigma-Aldrich Co. LLC
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Do not autoclave! Clean surface with 70% isopropyl alcohol.
Magnets should be kept away from magnetic storage devices, such as hard disks, ID cards, and credit cards, and from electronic devices and ferromagnetic materials. Persons with cardiac pacemakers should not work with these magnets.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library