Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0262

Sigma-Aldrich

Z-Phe-Ala-OH

≥99% (TLC), powder

Synonym(s):

Z-Phe-Ala, N-Cbz-Phe-Ala, Z-L-phenylalanyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2[OCH2CH(CH3)]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
Molecular Weight:
370.40
Beilstein:
4210469
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.32

product name

Z-Phe-Ala-OH,

Assay

≥99% (TLC)

Quality Level

form

powder

mol wt

370.41

solubility

methanol: ~50 mg/mL, almost clear, almost colorless

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)OCc2ccccc2)C(O)=O

InChI

1S/C20H22N2O5/c1-14(19(24)25)21-18(23)17(12-15-8-4-2-5-9-15)22-20(26)27-13-16-10-6-3-7-11-16/h2-11,14,17H,12-13H2,1H3,(H,21,23)(H,22,26)(H,24,25)/t14-,17-/m0/s1

InChI key

LEJTXQOVOPDGHS-YOEHRIQHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cathepsin A substrate

Packaging

pack of 5g and 25g

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service