Skip to Content
MilliporeSigma
Z-Phe-Ala-OH

Z-Phe-Ala-OH

Z-Phe-Ala-OH
Synonym(s):
Z-Phe-Ala, N-Cbz-Phe-Ala, Z-L-phenylalanyl-L-alanine
Linear Formula:
CH3CH(NH2)CH2[OCH2CH(CH3)]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
Molecular Weight:
370.40

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing