Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0106

Sigma-Aldrich

Caspase Substrate, chromogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39N7O10
Molecular Weight:
621.64
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Val-Gln-Val-Asp-pNA

Application

Ac-VQVD-pNa (Acetyl-Val-Gln-Val-Asp-p-nitroanilide) is used as a chromogenic caspase substrate.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Fang et al.
Journal of molecular biology, 360(3), 654-666 (2006-06-20)
The molecular basis for the substrate specificity of human caspase-3 has been investigated using peptide analog inhibitors and substrates that vary at the P2, P3, and P5 positions. Crystal structures were determined of caspase-3 complexes with the substrate analogs at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service