Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0094

Sigma-Aldrich

Caspase Inhibitor

Synonym(s):

Ac-AEVD-CHO, caspase 6 inhibitor, caspase 8 inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30N4O9
Molecular Weight:
458.46
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Gene Information

Amino Acid Sequence

Ac-Ala-Glu-Val-Asp-al

General description

Currently, there are 13 caspases recognized in mammals. Caspases are classified as initiators and effectors. Caspase 8 belongs to the class of initiators. It possesses a prodomain at the terminal and contains the death effector domain which serves as protein-protein interaction motif for caspase 8. With respect to substrate specificity, caspase 8 is classified as a member of group III. It specifically recognizes the (I/L/V) EXD sequence on the substrate.

Application

Caspase Inhibitor has been used to study the specificity of the CIL-1 (desglucoanagalloside B) interaction with the caspase 3/7 system.

Biochem/physiol Actions

The peptide Ac-Ala-Glu-Val-Asp-al, or Ac-AEVD-CHO, is an inhibitor of Group III caspases. It acts upon caspases-6, 8, 9, and 10, with particular effectiveness on caspase-6 and caspase-8. The use of Ac-AEVD-CHO in several apotosis studies has been reported.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triterpene saponosides from Lysimachia ciliata differentially attenuate invasive potential of prostate cancer cells
Koczurkiewicz P, et al.
Chemico-Biological Interactions, 206(1), 6-17 (2013)
Basic Concepts of Molecular Pathology, 32-32 (2009)
Alessandra De Leo et al.
PLoS pathogens, 13(8), e1006596-e1006596 (2017-08-31)
Kaposi's sarcoma (KS)-associated herpesvirus (KSHV) is a human gammaherpesvirus recognized as the principal causative agent of KS and primary effusion lymphoma (PEL). KSHV establishes persistent latent infection in B lymphocytes where viral gene expression is restricted, in part, by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service