Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0086

Sigma-Aldrich

Caspase 3 Substrate (PKC), fluorogenic

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H38N6O12S1
Molecular Weight:
706.72
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Caspase 3 Substrate (PKC), fluorogenic,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Asp-Met-Gln-Asp-AMC

Application

Caspase 3, a cysteine protease activator of caspase 6 and caspase 7, metabolizes amyloid-β 4A precursor protein. Caspase 3 may be detected using a variety of chromogenic and fluorogenic peptide substrates built around the DxxD (asp-x-x-asp) sequences DMQD and DEVD such as Ac-DMQD-pNa (Acetyl-Asp-Met-Gln-Asp-p- nitroanalide), chromogenic; Ac-DMQD-AMC (Acetyl-Asp-Met-Gln-Asp- amino-4-methylcoumarin), fluorogenic and Ac-DEVD-4M2Na.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sriram Ravindran et al.
Journal of biochemical and molecular toxicology, 34(12), e22606-e22606 (2020-09-01)
One of the common negative impacts in the management of acute myocardial infarction is cognitive decline. Using the rat model of isoproterenol (ISO)-induced myocardial infarction, we assessed the cardioprotective effect of sodium thiosulfate (STS) and its influence on cognition. STS
R Datta et al.
Blood, 88(6), 1936-1943 (1996-09-15)
The response of human myeloid leukemia cells to treatment with 1-beta-arabinofuranosylcytosine (ara-C) includes the induction of apoptosis. Ara-C induced apoptosis is associated with proteolytic cleavage of poly(ADP-ribose) polymerase (PARP) and protein kinase C (PKC) delta. However, the signals involved in
H Shimizu et al.
The Journal of investigative dermatology, 112(5), 769-774 (1999-05-08)
Exposure of human keratinocyte HaCaT cells to ultraviolet B-irradiation induced apoptotic morphologic changes. In this study, we found that the ultraviolet B irradiation (0.25 J per cm2) induced phosphorylation of p38 mitogen-activated protein kinase and c-jun N-terminal protein kinase, and
Osman Akdemir et al.
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia, 15(6), 672-678 (2008-04-02)
We investigated the therapeutic efficacy of Ac-DMQD-CHO, a caspase-3 inhibitor, and functional recovery in spinal cord injury in a rat model. Thirty rats were randomized into three groups of 10 each. In groups 2 and 3, spinal cord trauma was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service