MilliporeSigma
  • Activation of p38 mitogen-activated protein kinase and caspases in UVB-induced apoptosis of human keratinocyte HaCaT cells.
Activation of p38 mitogen-activated protein kinase and caspases in UVB-induced apoptosis of human keratinocyte HaCaT cells.
The Journal of investigative dermatology (1999-05-08)
H Shimizu, Y Banno, N Sumi, T Naganawa, Y Kitajima, Y Nozawa
ABSTRACT
Exposure of human keratinocyte HaCaT cells to ultraviolet B-irradiation induced apoptotic morphologic changes. In this study, we found that the ultraviolet B irradiation (0.25 J per cm2) induced phosphorylation of p38 mitogen-activated protein kinase and c-jun N-terminal protein kinase, and also significant activation of caspase-3 (CPP32-like protease) and a small increase of caspase-1 (ICE-like protease) activity in the early stages of ultraviolet B-induced apoptosis. Pretreatments of the cells with a p38 mitogen-activated protein kinase inhibitor, SB203580, and a caspase-3 inhibitor, Ac-Asp-Met-Gln-Asp-1-aldehyde, suppressed the ultraviolet B irradiation-induced apoptosis by approximately 60% as estimated by nuclear staining and DNA laddering. Pretreatment with caspase-1 inhibitor, Ac-Tyr-Val-Lys-Asp-aldehyde was without effect. Ultraviolet B-induced caspase-3 activation resulted in cleavage of poly(ADP) ribose polymerase, which was abolished by the caspase-3 inhibitor. SB203580 pretreatment prevented activation of caspase-3 and caspase-1, and also suppressed the cleavage of poly(ADP) ribose polymerase. Neither ceramide generation nor sphingomyelinase activation (neutral and acid) was observed in the ultraviolet B-irradiated HaCaT cells. Also various antioxidants did not affect the caspase activation induced by ultraviolet B irradiation. These results indicated that activation of p38 mitogen-activated protein kinase upstream of caspases may play an important part in the apoptotic process of keratinocytes exposed to ultraviolet B irradiation.
Sigma-Aldrich
Caspase 3 Substrate
Sigma-Aldrich
Caspase 3 Substrate (PKC), fluorogenic
Sigma-Aldrich
Caspase 3 Substrate (PKC), chromogenic
Sigma-Aldrich
Caspase 3 Apopain Substrate