Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAE0180

Sigma-Aldrich

3CL, Mpro protease fluorogenic substrate peptide

AFC-Peptide substrate, ≥95% (HPLC), solid

Synonym(s):

3CL protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.83

product name

3CL, Mpro protease fluorogenic substrate peptide, AFC-Peptide substrate, lyophilized

Quality Level

Assay

≥95% (HPLC)

form

solid

shipped in

ambient

storage temp.

−20°C

General description

This fluorogenic peptide is a substrate for the Main protease from SARS-Cov2, 3CL protease (Mpro). This substrate allows measurement of the activity of 3CL protease using a standard plate reader or spectrophotometer. Proteolytic cleavage by 3CL protease will release the fluorescent AFC group from the peptide (sequence TSAVLQ). After release, an increase in fluoresence is obtained (Excitation = 400 nm, Emission = 505 nm).

Peptide: Thr-Ser-Ala-Val-Leu-Gln-AFC

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service