Skip to Content
MilliporeSigma

SML2877

SARS-CoV-2 main protease inhibitor 11a

≥90% (HPLC)

Synonym(s):

N-((S)-3-cyclohexyl-1-oxo-1-((S)-1-oxo-3-((S)-2-oxopyrrolidin-3-yl)propan-2-ylamino)propan-2-yl)-1H-indole-2-carboxamide, N-[(1S)-1-(Cyclohexylmethyl)-2-[[(1S)-1-formyl-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]amino]-2-oxoethyl]-1H-indole-2-carboxamide, SARS-CoV-2 3CLpro inhibitor 11a, SARS-CoV-2 Mpro inhibitor 11a

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H32N4O4
CAS Number:
Molecular Weight:
452.55
UNSPSC Code:
51111800
NACRES:
NA.54
Assay:
≥90% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@@H](CC1CCCCC1)C(N[C@@H](C[C@H]2C(NCC2)=O)C=O)=O)C3=CC4=CC=CC=C4N3

InChI key

HPKJGHVHQWJOOT-ZJOUEHCJSA-N

Biochem/physiol Actions

COVID-19 corona virus SARS-CoV-2 main protease (Mpro; 3CLpro) substrate site Cys145 covalent inhibitor with low cytotoxicity in Vero E6 cultures.
Inhibitor 11a is a COVID-19 corona virus SARS-CoV-2 inhibitor that covalently targets the main protease (Mpro; a.k.a. 3C-like protease or 3CLpro) substrate site Cys145 (IC50 = 53 nM; 10 min pre-incubation with 30 nM SARS-CoV-2 Mpro prior to the addition of 20 μM substrate). Inhibitor 11a effectively inhibits SARS-CoV-2 replication in Vero E6 cultures (IC50 = 0.53 μM 24 h post infection with MOI = 0.05; cytotoxicity CC50 >100 μM) with good pharmacokinetic properties in mice in vivo (5 mg/kg via i.p. or i.v.).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library