Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAE0097

Sigma-Aldrich

D-2-Hydroxyglutarate Dehydrogenase (D2HGDH) from Acidaminococcus fermentans

recombinant, expressed in E. coli, aqueous solution

Synonym(s):

D2HGDH, HGDH, L-2-hydroxyglutarate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
1.1. 99.2
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

aqueous solution

specific activity

≥1000 units/mg protein

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

D-2-Hydroxyglutarate Dehydrogenase (D2HGDH) is a member of the D-2-hydroxyacid NAD+ dependent dehydrogenase family of proteins. D2HGDH catalyzes the conversion of α-ketoglutarate (α--KG) to D-2-hydroxyglutarate (D2HG), coupled to the oxidation of NADH to NAD+ .

The crystal structure of D2HGDH from Acidaminococcus fermentans has been reported. D2HGDH from Acidaminococcus fermentans has been used in several enzymatic assays, such as:
  • A continuous spectrophotometric assay to measure the activity of aminotransferases, based on the transamination of a keto compound and L-glutamate, which yields a corresponding amino compound and 2-oxoglutarate.
  • Determination of D2HG levels in biological fluids such as serum, urine, cell culture supernatants, and cell or tissue lysates.
  • A coupled assay system to measure branched-chain amino acid aminotransferase activity.

Unit Definition

One unit of enzyme oxidizes 1 μmole of NADH to NAD+ coupled to the reduction of α-ketoglutarate to (D)-2-hydroxyglutarate per minute at 37°C at pH 8.0.

Preparation Note

This recombinant D2HGDH product is supplied as an aqueous solution in 20 mM Trizma® buffer, pH 7.5, with 150 mM NaCl, and 10% glycerol.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuejing Yu et al.
Analytical biochemistry, 431(2), 127-131 (2012-09-25)
A continuous general spectrophotometric assay for measuring the activity of aminotransferases has been developed. It is based on the transamination of a keto compound (amino acceptor) and l-glutamate (amino donor), yielding the corresponding amino compound and 2-oxoglutarate. The rate of
Berta M Martins et al.
The FEBS journal, 272(1), 269-281 (2005-01-07)
NAD(+)-dependent (R)-2-hydroxyglutarate dehydrogenase (HGDH) catalyses the reduction of 2-oxoglutarate to (R)-2-hydroxyglutarate and belongs to the d-2-hydroxyacid NAD(+)-dependent dehydrogenase (d-2-hydroxyacid dehydrogenase) protein family. Its crystal structure was determined by phase combination to 1.98 A resolution. Structure-function relationships obtained by the comparison
Jörg Balss et al.
Acta neuropathologica, 124(6), 883-891 (2012-11-03)
Levels of (D)-2-hydroxyglutarate [D2HG, (R)-2-hydroxyglutarate] are increased in some metabolic diseases and in neoplasms with mutations in the isocitrate dehydrogenase 1 (IDH1) and isocitrate dehydrogenase 2 (IDH2) genes. Determination of D2HG is of relevance to diagnosis and monitoring of disease.
Xuejing Yu et al.
The FEBS journal, 281(1), 391-400 (2013-11-12)
Branched-chain amino acid aminotransferase (BCAT) plays a key role in the biosynthesis of hydrophobic amino acids (such as leucine, isoleucine and valine), and its substrate spectrum has not been fully explored or exploited owing to the inescapable restrictions of previous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service