MilliporeSigma
  • The specificity and kinetic mechanism of branched-chain amino acid aminotransferase from Escherichia coli studied with a new improved coupled assay procedure and the enzyme's potential for biocatalysis.
The specificity and kinetic mechanism of branched-chain amino acid aminotransferase from Escherichia coli studied with a new improved coupled assay procedure and the enzyme's potential for biocatalysis.
The FEBS journal (2013-11-12)
Xuejing Yu, Xingguo Wang, Paul C Engel
ABSTRACT
Branched-chain amino acid aminotransferase (BCAT) plays a key role in the biosynthesis of hydrophobic amino acids (such as leucine, isoleucine and valine), and its substrate spectrum has not been fully explored or exploited owing to the inescapable restrictions of previous assays, which were mainly based on following the formation/consumption of the specific branched-chain substrates rather than the common amino group donor/acceptor. In our study, detailed measurements were made using a novel coupled assay, employing (R)-hydroxyglutarate dehydrogenase from Acidaminococcus fermentans as an auxiliary enzyme, to provide accurate and reliable kinetic constants. We show that Escherichia coli BCAT can be used for asymmetric synthesis of a range of non-natural amino acids such as l-norleucine, l-norvaline and l-neopentylglycine and compare the kinetic results with the results of molecular modelling. A full two-substrate steady-state kinetic study for several substrates yields results consistent with a bi-bi ping-pong mechanism, and detailed analysis of the kinetic constants indicates that, for good 2-oxoacid substrates, release of 2-oxoglutarate is much slower than release of the product amino acid during the transamination reaction. The latter is in fact rate-limiting under conditions of substrate saturation. Branched-chain amino acid aminotransferase EC 2.6.1.42; (R)-2-hydroxyglutarate dehydrogenase EC 1.1.99.2.
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Norleucine, suitable for amino acid analysis, BioReagent
Sigma-Aldrich
L-Norleucine, ≥98% (TLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
SAFC
L-Valine
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-α-Neopentylglycine, ≥98.0% (TLC)
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)
Sigma-Aldrich
DL-Norvaline
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Sigma-Aldrich
D-2-Hydroxyglutarate Dehydrogenase (D2HGDH) from Acidaminococcus fermentans, recombinant, expressed in E. coli, aqueous solution