Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAE0054

Sigma-Aldrich

Lipoprotein, High Density from human plasma, solution

Synonym(s):

HDL, High Density Lipoprotein, alpha-Lipoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.61

form

liquid

Quality Level

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

High density lipoprotein (HDL) serves as the transit agent for cholesterol away from blood, tissues, and organs of the body to the liver, where cholesterol breakdown occurs. This process ultimately helps to remove cholesterol from the arteries. Colloquially known as the "good cholesterol", HDL makes up 20%-30% of the total cholesterol in the body.

HDL has been posited to be an important mediator in reversing metabolic dysfunction and regressing atherosclerotic cardiovascular disease. HDL is used in cardiovascular research and the development of potential therapeutic approaches to cardiovascular risk management.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service