Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12667-M

Sigma-Aldrich

Human Serum Albumin

30% Aqueous Solution

Synonym(s):

Albumin, Human Serum, 30% Aqueous Solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Product Name

Albumin, Human Serum, 30% Aqueous Solution,

biological source

human

description

Merck USA index - 14, 8468

Assay

≥95% (agarose gel electrophoresis)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

General description

Human serum albumin (HSA) is a soluble, globular, and unglycosylated monomeric protein, which is abundantly found in the plasma. It contains three homologous domains, numbered I, II, and III. Each domain is divided into subdomains A and B comprising common structural motifs. The HSA gene is localized to the human chromosome 4q13.3.

Biochem/physiol Actions

Human serum albumin (HSA) acts as the most important carrier protein for fatty acids, thyroid hormones, and steroids. It serves as a supplement in cell culture medium for the generation of vaccines and pharmaceuticals. HSA acts as a valuable biomarker for diseases, such as severe acute graft-versus-host disease, cancer, and rheumatoid arthritis. It is clinically used to treat serious burn injuries, hemorrhagic shock, hypoproteinemia, and fetal erythroblastosis. HSA is used in biological applications including nano delivery of drugs, carrier of oxygen, and fusion of peptides.

Warning

Toxicity: Standard Handling (A)

Physical form

30% solution in 20 mM Tris-HCl, 0.1% NaN₃, pH 6-8.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang He et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(47), 19078-19083 (2011-11-02)
Human serum albumin (HSA) is widely used in clinical and cell culture applications. Conventional production of HSA from human blood is limited by the availability of blood donation and the high risk of viral transmission from donors. Here, we report
Gabriella Fanali et al.
Molecular aspects of medicine, 33(3), 209-290 (2012-01-11)
Human serum albumin (HSA), the most abundant protein in plasma, is a monomeric multi-domain macromolecule, representing the main determinant of plasma oncotic pressure and the main modulator of fluid distribution between body compartments. HSA displays an extraordinary ligand binding capacity
Angelica M Merlot et al.
Frontiers in physiology, 5, 299-299 (2014-08-28)
Serum albumin is a multi-functional protein that is able to bind and transport numerous endogenous and exogenous compounds. The development of albumin drug carriers is gaining increasing importance in the targeted delivery of cancer therapy, particularly as a result of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service