SAB5702956
Anti-Vinculin Antibody
rabbit monoclonal, 9I3Y8
Synonym(s):
CMD1W, CMH15, HEL114, MV, MVCL
About This Item
Recommended Products
Product Name
Anti-Vinculin Antibody, clone 9I3Y8, Rabbit Monoclonal,
biological source
rabbit
Quality Level
material
colorless
clone
9I3Y8, monoclonal
form
liquid
mol wt
124 kDa
species reactivity
rat, human, rat
concentration
1.00 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
DAKAVAGNISDPGLQKSFLDSGYRILGAVAKVREAFQPQEPDFPPPPPDLEQLRLTDELAPPKPPLPEGEVPPPRPPPPEEKDEEFPEQKAGEVINQPMMM
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... VCL(7414)
General description
Immunogen
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service