Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539430

Sigma-Aldrich

Cyclohexanone diethyl ketal

97%

Synonym(s):

1,1-Diethoxycyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4360 (lit.)

bp

76-78 °C/20 mmHg (lit.)

density

0.908 g/mL at 25 °C (lit.)

functional group

ether
ketal

SMILES string

CCOC1(CCCCC1)OCC

InChI

1S/C10H20O2/c1-3-11-10(12-4-2)8-6-5-7-9-10/h3-9H2,1-2H3

InChI key

MWUDABUKTZAZCX-UHFFFAOYSA-N

General description

Cyclohexanone diethyl ketal (1,1-Diethoxycyclohexane) reacts with 1,3,2-benzodioxaborole [catecholborane (CB)] to form the corresponding ether. It undergoes thermal elimination reaction in gas-phase to yield 1-ethoxycyclohexene and ethanol.

Application

Cyclohexanone diethyl ketal may be used in the preparation of 1-ethoxycyclohexene.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix Rosas et al.
The journal of physical chemistry. A, 116(2), 846-854 (2011-12-22)
The gas-phase thermal elimination of 2,2-diethoxypropane was found to give ethanol, acetone, and ethylene, while 1,1-diethoxycyclohexane yielded 1-ethoxycyclohexene and ethanol. The kinetics determinations were carried out, with the reaction vessels deactivated with allyl bromide, and the presence of the free
Ozonolysis-reductive amination of olefins.
Pollart KA and Miller RE.
The Journal of Organic Chemistry, 27(7), 2392-2394 (1962)
Catecholborane (1, 3, 2-benzodioxaborole). A versatile reducing agent.
Kabalka GW, et al.
The Journal of Organic Chemistry, 42(3), 512-517 (1977)
Reactions of Enol Ethers with Carbenes. V. Rearrangements of Dihalocyclopropanes Derived from Six-, Seven-, and Eight-Membered Cyclic Enol Ethers1.
Parham WE, et al.
Journal of the American Chemical Society, 87(2), 321-328 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service