Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5500007

Sigma-Aldrich

Anti-Androgen Receptor (C-term) antibody, Rabbit monoclonal

clone SP242, recombinant, expressed in proprietary host, affinity isolated antibody

Synonym(s):

Anti-AIS, Anti-AR8, Anti-DHTR, Anti-HUMARA, Anti-HYSP1, Anti-KD, Anti-NR3C4, Anti-SBMA, Anti-SMAX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP242, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

dog, pig, rat, rabbit, mouse

technique(s)

immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... AR(367)

General description

Androgen receptor (AR) is a receptor for the androgenic hormones testosterone or dihydrotestosterone and it is a member of the steroid superfamily of ligand-dependent transcription factors. In the absence of androgen, the unligated AR mainly locates in cytoplasm, but translocates to the nucleus upon binding to androgen. Truncated AR variants with loss of C-terminus are common in prostate cancer.

Immunogen

Synthetic peptide from the C-terminus of human androgen receptor protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei Zhao et al.
Asian journal of urology, 6(1), 109-113 (2019-02-19)
Small cell prostate carcinoma (SCPC) is a rare and highly malignant subtype of prostate cancer. SCPC frequently lacks androgen receptor (AR) and prostate-specific antigen (PSA) expression, and often responds poorly to androgen deprivation therapy (ADT). AR splice variant-7 (AR-V7) is
Racial Differences in Androgen Receptor (AR) and AR Splice Variants (AR-SVs) Expression in Treatment-Na??ve Androgen-Dependent Prostate Cancer.
Khan, et al.
Biomedicines, 11 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service