Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5500006

Sigma-Aldrich

Anti-Androgen Receptor antibody, Rabbit monoclonal

clone SP107, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP107, monoclonal

species reactivity

human (tested)

species reactivity (predicted by homology)

dog, rat, mouse, pig

technique(s)

immunohistochemistry: 1:400

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... AR(367)

General description

The androgen receptor (AR) is a member of the steroid superfamily of ligand-dependent transcription factors. The expression of AR is reportedly inversely correlated with histologic grade, i.e., well differentiated prostate tumors show higher expression than the poorly differentiated tumors.

Immunogen

Synthetic peptide derived from near N-terminus of human AR.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paige M Glumac et al.
The Prostate, 78(13), 981-991 (2018-05-24)
The transmembrane glycoprotein CD133 is believed to be a marker of adult prostate stem cells and cancer stem/initiating cells. Investigating the role of CD133 in the normal biology of the prostate and in cancer is complicated by the lack of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service