Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5300151

Sigma-Aldrich

Monoclonal Anti-AIFM1 antibody produced in mouse

clone 4E7, ascites fluid

Synonym(s):

AIF, AIFM1, COXPD6, MGC111425, PDCD8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4E7, monoclonal

mol wt

67 kDa

species reactivity

mouse, monkey, human, rat

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG2b

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AIF(9131)

General description

AIFM1 (apoptosis inducing factor mitochondria associated 1) codes for apoptosis-inducing factor 1(AIF). It is located on human chromosome Xq26.1. AIF is a caspase-independent cell death effector.

Immunogen

Purified recombinant fragment of human AIF expressed in E.coli.
Mouse monoclonal antibody raised against AIF

Biochem/physiol Actions

AIFM1 (apoptosis inducing factor mitochondria associated 1) possesses redox activity, which helps in oxidative phosphorylation. In the mitochondria, AIFM1 helps in assembling or maintaining the mitochondrial respiratory complexes I and III. It plays a vital role in apoptosis. This gene is associated with COXPD6 (combined oxidative phosphorylation deficiency, encephalomyopathy, Cowchock syndrome and X-linked deafness with neuropathy.

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influenza virus infection induces translocation of apoptosis-inducing factor (AIF) in A549 cells: role of AIF in apoptosis and viral propagation.
Qu X, et al.
Archives of Virology, 162(3), 669-675 (2017)
Spondyloepimetaphyseal dysplasia with neurodegeneration associated with AIFM1 mutation?a novel phenotype of the mitochondrial disease.
Mierzewska H, et al.
Clinical Genetics, 91(1), 30-37 (2017)
X-linked hypomyelination with spondylometaphyseal dysplasia (H-SMD) associated with mutations in AIFM1.
Noriko M, et al.
Neurogenetics, 18(4), 185?194-185?194 (2017)
A newly distal hereditary motor neuropathy caused by a rare AIFM1 mutation.
Sancho P, et al.
Neurogenetics, 18(4), 245-250 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service