Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700912

Sigma-Aldrich

Monoclonal Anti-Perforin antibody produced in mouse

clone dG9

Synonym(s):

HPLH2, P1, PFP, PRF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

dG9, monoclonal

form

buffered aqueous solution

species reactivity

human, bovine

concentration

1 mg/mL

technique(s)

flow cytometry: 6 μg/mL

accession no.

NM_005041

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRF1(5551)

Related Categories

General description

The mouse monoclonal antibody dG9 (also known as deltaG9) recognizes perforin, a 70 kDa protein expressed in cytoplasmic granules of cytotoxic T cells and NK cells.

Immunogen

purified granules from human YT lymphoma cell line

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service