Skip to Content
MilliporeSigma
All Photos(1)

Documents

SAB4700750

Sigma-Aldrich

Monoclonal Anti-CD117 antibody produced in mouse

clone 104D2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-c-Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

104D2, monoclonal

form

buffered aqueous solution

species reactivity

bovine, nonhuman primates, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)

General description

The mouse monoclonal antibody 104D2 detects extracellular part of CD117 / c-Kit protooncogen.
HLDA VI; WS Code C-30.

Immunogen

MOLM-1 megakaryocytic cells

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weina Zhang et al.
Frontiers of medicine, 13(3), 388-397 (2018-09-13)
Collaboration of c-KIT mutations with AML1-ETO (AE) has been demonstrated to induce t(8; 21) acute myeloid leukemia (AML). Targeted therapies designed to eliminate AE and c-KIT oncoproteins may facilitate effective treatment of t(8; 21) AML. Homoharringtonine (HHT) features activity against
Bao Yang et al.
Oncology letters, 14(3), 3825-3831 (2017-09-21)
Myeloid sarcoma (MS) is a localized tumor composed of premature precursors of granulocytic cells, which may occur in any organ and most commonly involves the soft tissue and musculoskeletal system. This malignancy may occur in the presence or absence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service