Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700729

Sigma-Aldrich

IgG2b Isotype Control murine

clone MPC-11, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.42

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

MPC-11, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

shipped in

wet ice

storage temp.

2-8°C

General description

This mouse IgG2b (kappa) monoclonal antibody (clone MPC-11) reacts with an epitope irrelevant for a variety of resting, activated, live, and fixed human, mouse, and rat tissues.
The specificity of staining by monoclonal antibodies to target antigens should be verified by establishing the amount of non-specific antibody binding. Especially at higher concentration (more than 15 μg/ml) the antibody staining usually has consignable background. To this end a non-reactive immunoglobulin of the same isotype is included as a negative control for each specific monoclonal antibody used in a particular immunoassay. The monoclonal antibody MPC-11 was generated against an epitope irrelevant for human, mouse, and rat material, and can thus be used for evaluation of the background staining that is caused by general nonspecific interactions between an mouse IgG2b molecule and the respective sample under the particular conditions.

Immunogen

KLH-coupled trinitrophenol

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Steinle et al.
The Journal of experimental medicine, 181(2), 503-513 (1995-02-01)
The nature of alloantigens seen by T lymphocytes, in particular the role of peptides in allorecognition, has been studied intensively whereas knowledge about the in vivo emergence, diversity, and the structural basis of specificity of alloreactive T cells is very
Rosanna C G Smith et al.
Biophysical journal, 112(12), 2641-2652 (2017-06-22)
A number of important pluripotency regulators, including the transcription factor Nanog, are observed to fluctuate stochastically in individual embryonic stem cells. By transiently priming cells for commitment to different lineages, these fluctuations are thought to be important to the maintenance
George C Brittain et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 91(4), 351-363 (2017-04-04)
A biochemical system and method have been developed to enable the quantitative measurement of cytoplasmic versus nuclear localization within cells in whole blood. Compared with the analyses of nuclear localization by western blot or fluorescence microscopy, this system saves a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service